Merck
  • Measurement of nitrosamine and nitramine formation from NOx reactions with amines during amine-based carbon dioxide capture for postcombustion carbon sequestration.
Measurement of nitrosamine and nitramine formation from NOx reactions with amines during amine-based carbon dioxide capture for postcombustion carbon sequestration.
Environmental science & technology (2012-07-27)
Ning Dai, Amisha D Shah, Lanhua Hu, Michael J Plewa, Bruce McKague, William A Mitch
ABSTRACT
With years of full-scale experience for precombustion CO(2) capture, amine-based technologies are emerging as the prime contender for postcombustion CO(2) capture. However, concerns for postcombustion applications have focused on the possible contamination of air or drinking water supplies downwind by potentially carcinogenic N-nitrosamines and N-nitramines released following their formation by NO(x) reactions with amines within the capture unit. Analytical methods for N-nitrosamines in drinking waters were adapted to measure specific N-nitrosamines and N-nitramines and total N-nitrosamines in solvent and washwater samples. The high levels of amines, aldehydes, and nitrite in these samples presented a risk for the artifactual formation of N-nitrosamines during sample storage or analysis. Application of a 30-fold molar excess of sulfamic acid to nitrite at pH 2 destroyed nitrite with no significant risk of artifactual nitrosation of amines. Analysis of aqueous morpholine solutions purged with different gas-phase NO and NO(2) concentrations indicated that N-nitrosamine formation generally exceeds N-nitramine formation. The total N-nitrosamine formation rate was at least an order of magnitude higher for the secondary amine piperazine (PZ) than for the primary amines 2-amino-2-methyl-1-propanol (AMP) and monoethanolamine (MEA) and the tertiary amine methyldiethanolamine (MDEA). Analysis of pilot washwater samples indicated a 59 μM total N-nitrosamine concentration for a system operated with a 25% AMP/15% PZ solvent, but only 0.73 μM for a 35% MEA solvent. Unfortunately, a greater fraction of the total N-nitrosamine signal was uncharacterized for the MEA-associated washwater. At a 0.73 μM total N-nitrosamine concentration, a ~25000-fold reduction in concentration is needed between washwater units and downwind drinking water supplies to meet proposed permit limits.
Sigma-Aldrich
Sulfamic acid, JIS special grade, ≥99.5%
Supelco
Sulfamic acid, analytical standard (for acidimetry), ACS reagent
Sigma-Aldrich
Sulfamic acid, 99.999% trace metals basis
Sigma-Aldrich
Ammonium sulfamate, JIS special grade, ≥98.5%
Sigma-Aldrich
Sulfamic acid, ≥99.5% (alkalimetric)
Sigma-Aldrich
Ammonium sulfamate, BioXtra, ≥98.0%
Sigma-Aldrich
Sulfamic acid, reagent grade, 98%
Sigma-Aldrich
Ammonium sulfamate, ACS reagent, ≥98.0%
Sigma-Aldrich
Sulfamic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Sulfamic acid, ACS reagent, 99.3%