Merck
  • The role of the alcohol and carboxylic acid in directed ruthenium-catalyzed C(sp3)-H α-alkylation of cyclic amines.
The role of the alcohol and carboxylic acid in directed ruthenium-catalyzed C(sp3)-H α-alkylation of cyclic amines.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-07-13)
Sheba D Bergman, Thomas E Storr, Hana Prokopcová, Karel Aelvoet, Gaston Diels, Lieven Meerpoel, Bert U W Maes
ABSTRACT
A general directed Ru-catalyzed C(sp(3))-H α-alkylation protocol for piperidines (less-reactive substrates than the corresponding five-membered cyclic amines) has been developed. The use of a hindered alcohol (2,4-dimethyl-3-pentanol) as the solvent and catalyst activator, and a catalytic amount of trans-1,2-cyclohexanedicarboxylic acid is necessary to achieve a high conversion to product. This protocol was used to effectively synthesize a number of 2-hexyl- and 2,6-dihexyl piperidines, as well as the alkaloid (±)-solenopsin A. Kinetic studies have revealed that the carboxylic acid additive has a significant effect on catalyst initiation, catalyst longevity, and reverses the reaction selectivity compared with the acid-free reaction (promotes alkylation versus competing alkene reduction).
Sigma-Aldrich
1-Propanol, JIS special grade
Supelco
1-Propanol, analytical standard
Sigma-Aldrich
1-Propanol, anhydrous, 99.7%
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
1-Propanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
1-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
1-Propanol, ≥99% (GC), purum
Sigma-Aldrich
1-Propanol, suitable for HPLC, ≥99.9%