Merck
  • Surface treatment of hematite photoanodes with zinc acetate for water oxidation.
Surface treatment of hematite photoanodes with zinc acetate for water oxidation.
Nanoscale (2012-06-13)
Lifei Xi, Prince Saurabh Bassi, Sing Yang Chiam, Wai Fatt Mak, Phong D Tran, James Barber, Joachim Say Chye Loo, Lydia Helena Wong
ABSTRACT
A simple and inexpensive method to form a hematite photoanode for efficient water oxidation is reported. A very thin ZnO overlayer was deposited on top of a thin film of hematite and found, compared with non-treated hematite, to increase the photocurrent and reduce the onset potential for generating oxygen from water. After 3 cycles of ZnAc treatment, the photocurrent increased more than 40% to 1.08 mA cm(-2) at 0.23 V vs. Ag/AgCl and onset potential for water oxidation shifted by -170 mV. It is proposed that the ZnO overlayer changes the flat band potential of hematite and reduces the surface defects.
Sigma-Aldrich
Zinc acetate dihydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Zinc acetate dihydrate, 99.999% trace metals basis
Sigma-Aldrich
Zinc acetate dihydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinc acetate dihydrate, reagent grade
Sigma-Aldrich
Zinc acetate dihydrate, puriss., E 650, 99-102%
Sigma-Aldrich
Zinc acetate dihydrate, ACS reagent, ≥98%