Merck
  • Novel method for the determination of the methoxyl content in lignin by headspace gas chromatography.
Novel method for the determination of the methoxyl content in lignin by headspace gas chromatography.
Journal of agricultural and food chemistry (2012-05-15)
Hailong Li, Xin-Sheng Chai, Mengru Liu, Yonghong Deng
ABSTRACT
The paper reports on a headspace gas chromatographic (HS-GC) method for the determination of methoxyl in lignin. The method involves the quantitive cleavage of methoxyl with hydroiodic acid (HI) to form methyl iodide in a closed headspace sample vial at 130 °C for 30 min. After HI has been added, the sample is neutralized by injecting a sodium hydroxide solution; the methyl iodide in the vial was determined by HS-GC using a flame ionization detector. The results showed that the method has an excellent measurement precision (RSD < 0.69%) and accuracy (RSD < 3.5%) for the quantification of methoxyl content in lignin. The present method is simple and accurate and can be used for the efficient determination of methoxy1 content in lignin and related materials.
Sigma-Aldrich
Hydriodic acid, 57 wt. %, distilled, 99.999% trace metals basis
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, distilled, 57 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Hydriodic acid, 57 wt. % in H2O, distilled, stabilized, 99.95%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, ACS reagent, 55%
Sigma-Aldrich
Hydriodic acid, contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%