Merck
  • Oral submucous fibrosis: a report of two pediatric cases and a brief review.
Oral submucous fibrosis: a report of two pediatric cases and a brief review.
Journal of the Indian Society of Pedodontics and Preventive Dentistry (2012-05-09)
R Dhariwal, J G Ray, S Mohanty Pattanayak, N Swain
ABSTRACT
Oral submucous fibrosis is a chronic debilitating disorder which is usually seen in adults with areca nut chewing habit. A rapid upsurge in the popularity of commercial areca nut products, especially among the younger generation, is a cause of grave concern, as satisfactory management is still lacking. Hereby, two pediatric cases of oral submucous fibrosis are reported.
Sigma-Aldrich
Zinc acetate dihydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Zinc acetate dihydrate, 99.999% trace metals basis
Sigma-Aldrich
Zinc acetate dihydrate, reagent grade
Sigma-Aldrich
Zinc acetate dihydrate, ACS reagent, ≥98%
Sigma-Aldrich
Zinc acetate dihydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinc acetate dihydrate, puriss., E 650, 99-102%