Merck
  • Selective reduction of biomass by hydriodic acid and its in situ regeneration from iodine by metal/hydrogen.
Selective reduction of biomass by hydriodic acid and its in situ regeneration from iodine by metal/hydrogen.
ChemSusChem (2012-04-12)
Weiran Yang, Matthew R Grochowski, Ayusman Sen
ABSTRACT
HI returns: Hydroiodic acid is a highly selective reducing reagent for a wide variety of substrates. Its application is limited by the formation of iodine and the difficulty in reconverting that iodione back to HI in situ. We report the facile conversion of I(2) to HI by metal-catalyzed hydrogenation in the presence of water, and demonstrate the utility of this process in the conversion of fructose to 5-methyfurfural and glycerol to 2-iodopropane.
Sigma-Aldrich
Hydriodic acid, contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%
Sigma-Aldrich
Hydriodic acid, 57 wt. % in H2O, distilled, stabilized, 99.95%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, distilled, 57 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, ACS reagent, 55%
Sigma-Aldrich
Hydriodic acid, 57 wt. %, distilled, 99.999% trace metals basis