Merck
  • Essential oil of Myrica esculenta Buch. Ham.: composition, antimicrobial and topical anti-inflammatory activities.
Essential oil of Myrica esculenta Buch. Ham.: composition, antimicrobial and topical anti-inflammatory activities.
Natural product research (2012-01-21)
Supriya Agnihotri, Sharad Wakode, M Ali
ABSTRACT
Hydrodistilled oil obtained from the stem bark of Myrica esculenta Buch. Ham. ex D. Don (yield 0.3%) was analysed by capillary GC and GC-MS. The volatile oil consisted mainly of n-hexadecanol (25.2%), eudesmol acetate (21.9%), palmitic acid (11.6%), cis-β-caryophyllene (8.7%), n-pentadecanol (7.7%) and n-octadecanol (7.6%). The oil was found to be a potential antimicrobial agent against Bacillus pumilus, Staphylococcus aureus, Staphylococcus epidermidis, Escherichia coli, Pseudomonas aeruginosa, Candida albicans, Aspergillus niger and Saccharomyces cerevisiae. The essential oil exhibited significant topical anti-inflammatory activity compared to standard drug in Swiss albino mice ear.
Product Description
Stearyl alcohol, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Hexadecanol, 95%
Sigma-Aldrich
1-Hexadecanol, ReagentPlus®, 99%
Sigma-Aldrich
4-Allyl-2,6-dimethoxyphenol, ≥95%, FG
Sigma-Aldrich
1-Hexadecanol, ≥99%
Sigma-Aldrich
4-Allyl-2,6-dimethoxyphenol, ≥95%
Sigma-Aldrich
Cetyl alcohol, SAJ special grade, ≥98.0%
Supelco
Cetyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
1-Octadecanol, Selectophore, ≥99.0%
Sigma-Aldrich
1-Octadecanol, ReagentPlus®, 99%
Supelco
Cetyl alcohol, analytical standard
Sigma-Aldrich
1-Octadecanol, 95%
Cetyl alcohol, European Pharmacopoeia (EP) Reference Standard