Merck
  • Bromocresol green/mesoporous silica adsorbent for ammonia gas sensing via an optical sensing instrument.
Bromocresol green/mesoporous silica adsorbent for ammonia gas sensing via an optical sensing instrument.
Sensors (Basel, Switzerland) (2011-12-14)
Yu-Chang Chang, Hsunling Bai, Shou-Nan Li, Chun-Nan Kuo
ABSTRACT
A meso-structured Al-MCM-41 material was impregnated with bromocresol green (BG) dye and then incorporated into a UV-Vis DRA spectroscopic instrument for the online detection of ammonia gas. The absorption response of the Al-MCM-41/BG ammonia sensing material was very sensitive at the optical absorption wavelength of 630 nm. A high linear correlation was achieved for ppmv and sub-ppmv levels of ammonia gas. The response time for the quantitative detection of ammonia gas concentrations ranging from 0.25 to 2.0 ppmv was only a few minutes. The lower detection limit achieved was 0.185 ppmv. The color change process was fully reversible during tens of cycling tests. These features together make this mesoporous Al-MCM-41 material very promising for optical sensing applications.
Sigma-Aldrich
Bromocresol Green, JIS special grade
Sigma-Aldrich
Bromocresol Green, ACS reagent, Dye content 95 %
Sigma-Aldrich
Bromocresol Green solution, pH 3.8-5.4
Sigma-Aldrich
Bromocresol Green Sultone Form, for microscopy (Bot., Hist., Vit.), indicator (pH 3.8-5.4)