Merck
Sonochemical proteinaceous microspheres for wound healing.
Advances in experimental medicine and biology (2011-11-22)
Raquel Silva, Helena Ferreira, Andreia Vasconcelos, Andreia C Gomes, Artur Cavaco-Paulo
ABSTRACT
In this work, we report a novel approach using proteinaceous microspheres of bovine serum albumin (BSA), human serum albumin (HSA) and silk fibroin (SF) containing different organic solvents, namely n-dodecane, mineral oil and vegetable oil, to reduce the activity of human neutrophil elastase (HNE) found in high levels on chronic wounds. The ability of these devices to inhibit HNE was evaluated using porcine pancreatic elastase (PPE) solution as a model of wound exudates. The results obtained indicated that the level of PPE activity can be tuned by changing the organic solvent present on different protein microspheres, thus showing an innovative way of controlling the elastase-antielastase imbalance found in chronic wounds. Furthermore, these proteinaceous microspheres were shown to be important carriers of elastase inhibitors causing no cytotoxicity in human skin fibroblasts in vitro, making them suitable for biomedical applications, such as chronic wounds.
Product Description
Supelco
Dodecane, analytical standard
Sigma-Aldrich
Dodecane, anhydrous, ≥99%
Supelco
Density Standard 749 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Dodecane, ReagentPlus®, ≥99%