Merck
  • Plastic bag method for active sample loading into transmission electron microscope.
Plastic bag method for active sample loading into transmission electron microscope.
Journal of electron microscopy (2011-10-05)
Hao Yao, Shigehito Isobe, Yongming Wang, Naoyuki Hashimoto, Somei Ohnuki
ABSTRACT
A plastic bag method was developed to observe air-sensitive samples on microstructure and phase distribution without exposure to air during the holder transfer process into the transmission electron microscope (TEM). As an example, a type of lithium aluminum hydride (Li(3)AlH(6)) was observed in the TEM to demonstrate the effectiveness of this method. Results show that the plastic bag method is a simple and practical TEM transfer method utilized to reduce air contact for a series of air-sensitive materials.
Sigma-Aldrich
Lithium aluminum hydride solution, 2.3 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Lithium aluminum hydride, hydrogen-storage grade
Sigma-Aldrich
Lithium aluminum hydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride, pellets, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric)
Sigma-Aldrich
Lithium aluminum hydride, powder, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride solution, 2.0 M in THF