Merck
  • Mass transport at rotating disk electrodes: effects of synthetic particles and nerve endings.
Mass transport at rotating disk electrodes: effects of synthetic particles and nerve endings.
Analytical biochemistry (2011-08-02)
Veronica M Chiu, Peter A Lukus, Jamie L Doyle, James O Schenk
ABSTRACT
An unstirred layer (USL) exists at the interface of solids with solutions. Thus, the particles in brain tissue preparations possess a USL as well as at the surface of a rotating disk electrode (RDE) used to measure chemical fluxes. Time constraints for observing biological kinetics based on estimated thicknesses of USLs at the membrane surface in real samples of nerve endings were estimated. Liposomes, silica, and Sephadex were used separately to model the tissue preparation particles. Within a solution stirred by the RDE, both diffusion and hydrodynamic boundary layers are formed. It was observed that the number and size of particles decreased the following: the apparent diffusion coefficient excluding Sephadex, boundary layer thicknesses excluding silica, sensitivity excluding diluted liposomes (in agreement with results from other laboratories), limiting current potentially due to an increase in the path distance, and mixing time. They have no effect on the detection limit (6 ± 2 nM). The RDE kinetically resolves transmembrane transport with a timing of approximately 30 ms.
Sigma-Aldrich
Sephadex® G-10, Medium
Sigma-Aldrich
Sephadex® G-50, Superfine
Sigma-Aldrich
Sephadex® G-50, BioReagent, DNA grade, for molecular biology, medium
Supelco
Sephadex® G-50, Medium
Sigma-Aldrich
Sephadex® G-50, Fine
Sigma-Aldrich
Sephadex® G-50, BioReagent, DNA grade, for molecular biology, fine
Sigma-Aldrich
Sephadex® G-15, Medium
Sigma-Aldrich
Sephadex® G-100, Superfine
Sigma-Aldrich
Sephadex® G-25, Medium
Sigma-Aldrich
Sephadex® G-75, Superfine
Sigma-Aldrich
Sephadex® G-25, Fine
Sigma-Aldrich
Sephadex® G-100, Medium
Sigma-Aldrich
Sephadex® G-75, Medium
Sigma-Aldrich
Sephadex® G-25, DNA grade, BioReagent, for molecular biology, Superfine
Sigma-Aldrich
Sephadex® G-25, Coarse
Sigma-Aldrich
Sephadex® G-100, BioReagent, DNA grade, for molecular biology, medium