Merck
  • Carbonaceous nanofiber membrane functionalized by beta-cyclodextrins for molecular filtration.
Carbonaceous nanofiber membrane functionalized by beta-cyclodextrins for molecular filtration.
ACS nano (2011-06-15)
Ping Chen, Hai-Wei Liang, Xiao-Han Lv, Hai-Zhou Zhu, Hong-Bin Yao, Shu-Hong Yu
ABSTRACT
In this paper, we report the fabrication of carbonaceous nanofiber (CNF) membranes functionalized by beta-cyclodextrins (CNF-β-CD membrane) and their application for molecular filtration. The chemically synthesized carbonaceous nanofibers were first functionalized by β-CD, and the free-standing CNF membrane can be prepared by a simple filtration process. The membrane shows a remarkable capability to function as an ideal molecular filter through complexation of phenolphthalein molecules with the cyclodextrin molecules grafted on the CNFs. As a typical dye pollutant, fuchsin acid can also be effectively removed from the solution through such a membrane. Engineering the surface of this carbonaceous nanofiber membrane may allow it to be used for other applications such as chiral separation and drug delivery.
Sigma-Aldrich
Phenolphthalein solution, 0.1 % (w/v), pH 7.8-10.0
Sigma-Aldrich
Phenolphthalein solution, 0.04 % (w/v), pH 7.8-10.0
Sigma-Aldrich
Phenolphthalein, JIS special grade, ≥98.0%
Sigma-Aldrich
Phenolphthalein, SAJ first grade, ≥97.0%
Sigma-Aldrich
Phenolphthalein solution, 1 % (w/v), pH 7.8-10.0
Sigma-Aldrich
Phenolphthalein solution, 0.5 wt. % in ethanol: water (1:1)
Sigma-Aldrich
Phenolphthalein, ACS reagent