Merck
  • Different solvents for the regeneration of the exhausted activated carbon used in the treatment of coking wastewater.
Different solvents for the regeneration of the exhausted activated carbon used in the treatment of coking wastewater.
Journal of hazardous materials (2011-01-18)
Dongsheng Guo, Qiantao Shi, Binbin He, Xiaoying Yuan
ABSTRACT
The solvents n-pentane, methylene dichloride, ethyl ether and dodecylbenzenesulphonic acid sodium were used to regenerate exhausted activated carbon used in the process of treating coking wastewater, and the efficiency, ability, and optimum conditions of the different solvents on this regeneration were investigated. The results indicate that n-pentane could effectively remove refractory organic compounds in the coking wastewater adsorbed on the surface of activated carbon and could repeatedly regenerate the exhausted activated carbon to recover its adsorption activity. Under the conditions of a regeneration time of 20 min, a regeneration temperature of 25°C, an activated carbon drying time of 300 min, and an activated carbon drying temperature of 150°C, n-pentane had the best regeneration efficiency, at 98.27%, for exhausted activated carbon. Gas chromatography-mass spectrometry analysis results show that the nature of the activated carbon regenerated by organic solvents had no remarkable change in adsorption for the main types of organic compounds in coking wastewater. The good regenerative effect of n-pentane on the activated carbon may be due its stronger desorption of esters embedded within the internal structure of activated carbon.
Sigma-Aldrich
Pentane, SAJ special grade, ≥99.0%
Sigma-Aldrich
Pentane, SAJ first grade, ≥96.0%
Sigma-Aldrich
Pentane, anhydrous, ≥99%
Supelco
Pentane, analytical standard
Sigma-Aldrich
Pentane, ≥99% (GC)
Sigma-Aldrich
Pentane, reagent grade, 98%
Sigma-Aldrich
Pentane, spectrophotometric grade, ≥99%
Sigma-Aldrich
Pentane, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Pentane, suitable for HPLC, ≥99.0%