Merck
  • Evaluating the impact of water supply strategies on p-xylene biodegradation performance in an organic media-based biofilter.
Evaluating the impact of water supply strategies on p-xylene biodegradation performance in an organic media-based biofilter.
Journal of hazardous materials (2010-10-30)
G Gallastegui, R Muñoz, A Barona, G Ibarra-Berastegi, N Rojo, A Elías
ABSTRACT
The influence of water irrigation on both the long-term and short-term performance of p-xylene biodegradation under several organic loading scenarios was investigated using an organic packing material composed of pelletised sawdust and pig manure. Process operation in a modular biofilter, using no external water supply other than the moisture from the saturated inlet air stream, showed poor p-xylene abatement efficiencies (≈33 ± 7%), while sustained irrigation every 25 days rendered a high removal efficiency (RE) for a critical loading rate of 120 g m(-3)h(-1). Periodic profiles of removal efficiency, temperature and moisture content were recorded throughout the biofilter column subsequent to each biofilter irrigation. Hence, higher p-xylene biodegradation rates were always initially recorded in the upper module, which resulted in a subsequent increase in temperature and a decrease in moisture content. This decrease in the moisture content in the upper module resulted in a higher removal rate in the middle module, while the moisture level in the lower module steadily increased as a result of water condensation. Based on these results, mass balance calculations performed using measured bed temperatures and relatively humidity values were successfully used to account for water balances in the biofilter over time. Finally, the absence of bed compaction after 550 days of continuous operation confirmed the suitability of this organic material for biofiltration processes.
Sigma-Aldrich
p-Xylene, SAJ first grade, ≥99.0%
Supelco
p-Xylene, analytical standard
Sigma-Aldrich
p-Xylene, anhydrous, ≥99%
Supelco
p-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
p-Xylene, ReagentPlus®, 99%
Sigma-Aldrich
p-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
p-Xylene, suitable for HPLC, ≥99%