Merck
[(Methyl)1-(11)c]-acetate metabolism in hepatocellular carcinoma.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging (2010-04-20)
Nicolas Salem, Yu Kuang, David Corn, Bernadette Erokwu, Jeffrey A Kolthammer, Haibin Tian, Chunying Wu, Fangjing Wang, Yanming Wang, Zhenghong Lee
ABSTRACT
Studies have established the value of [(methyl)1-(11)C]-acetate ([(11)C]Act) combined with 2-deoxy-2[(18)F]fluoro-D-glucose (FDG) for detecting hepatocellular carcinoma (HCC) using positron emission tomography (PET). In this study, the metabolic fate of [(11)C]Act in HCC was characterized. Experiments with acetic acid [1-(14)C] sodium salt ([(14)C]Act) were carried out on WCH-17 cells and freshly derived rat hepatocytes. PET scans with [(11)C]Act were also carried out on woodchucks with HCC before injection of [(14)C]Act. The radioactivity levels in different metabolites were quantified with thin-layer chromatography. In WCH-17 cells, the predominant metabolite was phosphatidylcholine (PC). Regions of HCCs with the highest [(11)C]Act uptake had higher radioactivity accumulation in lipid-soluble compounds than surrounding hepatic tissues. In those regions, PC and triacylglycerol (TG) accumulated more radioactivity than in surrounding hepatic tissues. High [(11)C]Act uptake in HCC is associated with increased de novo lipogenesis. PC and TG are the main metabolites into which the radioactive label from [(11)C]Act is incorporated in HCC.
Sigma-Aldrich
Methyl acetate, JIS special grade, ≥99.5%
Sigma-Aldrich
Methyl acetate, natural, 98%, FG
Supelco
Methyl acetate, analytical standard
Sigma-Aldrich
Methyl acetate, ≥98%, FG
Sigma-Aldrich
Methyl acetate, anhydrous, 99.5%
Sigma-Aldrich
Methyl acetate, ReagentPlus®, 99%