Merck
  • Scanning tunneling microscopy investigation of self-assembled CuPc/F16CuPc binary superstructures on graphite.
Scanning tunneling microscopy investigation of self-assembled CuPc/F16CuPc binary superstructures on graphite.
Langmuir : the ACS journal of surfaces and colloids (2010-02-24)
Yu Li Huang, Hui Li, Jing Ma, Han Huang, Wei Chen, Andrew T S Wee
ABSTRACT
The self-assembly of the binary molecular system comprising copper(II) phthalocyanine (CuPc) and copper-hexadecafluoro-phthalocyanine (F(16)CuPc) on graphite has been investigated by in situ low-temperature scanning tunneling microscopy (LT-STM). The adsorption of this binary molecular system on graphite results in the formation of a well-ordered chessboardlike nanopattern. The in-plane molecular orientation of the guest CuPc molecules can be tuned by varying the coverage. At low coverage, the sparse CuPc molecules are randomly embedded in the host F(16)CuPc monolayer, possessing two different in-plane orientations; as the CuPc coverage increases, the in-plane molecular orientations of CuPc and F(16)CuPc become unidirectional and a highly ordered chessboardlike pattern forms. Molecular dynamic (MD) simulation results suggest that the selective and directional intermolecular hydrogen bonding determines the in-plane molecular orientation as well as the supramolecular packing arrangement.
Sigma-Aldrich
Copper(II) phthalocyanine, triple-sublimed grade, >99.95% trace metals basis
Sigma-Aldrich
Copper(II) phthalocyanine, Dye content >99 %
Sigma-Aldrich
Copper(II) phthalocyanine, sublimed grade, Dye content 99 %
Sigma-Aldrich
Copper(II) phthalocyanine, β-form, Dye content 90 %