Merck
  • Role of tyrosine 131 in the active site of paAzoR1, an azoreductase with specificity for the inflammatory bowel disease prodrug balsalazide.
Role of tyrosine 131 in the active site of paAzoR1, an azoreductase with specificity for the inflammatory bowel disease prodrug balsalazide.
Acta crystallographica. Section F, Structural biology and crystallization communications (2010-01-09)
Chan-Ju Wang, Nicola Laurieri, Areej Abuhammad, Edward Lowe, Isaac Westwood, Ali Ryan, Edith Sim
ABSTRACT
Azoreductase 1 from Pseudomonas aeruginosa strain PAO1 (paAzoR1) catalyses the activation of the prodrug balsalazide and reduces the azo dye methyl red using reduced nicotinamide adenine dinucleotide cofactor as an electron donor. To investigate the mechanism of the enzyme, a Y131F mutation was introduced and the enzymic properties of the mutant were compared with those of the wild-type enzyme. The crystallographic structure of the mutant with methyl red bound was solved at 2.1 A resolution and compared with the wild-type structure. Tyr131 is important in the architecture of the active site but is not essential for enzymic activity.
Sigma-Aldrich
Methyl Red solution, pH 4.2-6.2
Millipore
Methyl Red solution, suitable for microbiology
Sigma-Aldrich
Methyl Red, ACS reagent, crystalline
Sigma-Aldrich
Methyl Red, JIS special grade
Sigma-Aldrich
Methyl Red sodium salt, Crystalline
Sigma-Aldrich
Methyl Red sodium salt, ACS reagent, Dye content 95 %