Merck
  • Determination of creatine and guanidinoacetate by GC-MS: study of their stability in urine at different temperatures.
Determination of creatine and guanidinoacetate by GC-MS: study of their stability in urine at different temperatures.
Clinical biochemistry (2008-11-11)
José Angel Prieto, Fernando Andrade, Sergio Martín, Pablo Sanjurjo, Javier Elorz, Luís Aldámiz-Echevarría
ABSTRACT
Evaluation of a GC-MS method using N-tert-butyldimethylsilyl-N-methyltrifluoroacetamide (MTBSTFA) as the silylating agent for GC-MS. Study of the stability of creatine and guanidinoacetate in urine. 22 urines were kept at RT, 4 degrees C and -30 degrees C for 15 days. MTBSTFA produces a single chromatographic peak in contrast with other derivatizing agents. Creatine concentration increases at room temperature (326% on average), and at 4 degrees C (75%). However, detection decreases after freezing (-37%). Guanidinoacetate is stable, but decreases after freezing (-37%). Sonication before analysis is crucial to obtain repetitive results. A modified GC-MS method has been validated and the conditions for preservation of the urine have been established.
Product Description
Supelco
MTBSTFA (with 1% t-BDMCS), (with 1% t-BDMCS), ampule of 5 × 1 mL, analytical standard, Cerilliant®
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, ≥95%
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide, >97%