Merck
  • Improved surface-enhanced Raman scattering on electrochemically roughened silver substrates prepared in bielectrolyte solutions.
Improved surface-enhanced Raman scattering on electrochemically roughened silver substrates prepared in bielectrolyte solutions.
Analytica chimica acta (2007-03-28)
Yu-Chuan Liu, Cheng-Cai Wang, Jeng-Feng Tsai
ABSTRACT
In this work, the effect of supplemental LiClO(4) electrolytes in KCl solutions used in roughening silver substrates by electrochemical triangular-wave oxidation-reduction cycles (ORC) on surface-enhanced Raman scattering (SERS) was first investigated. To prepare SERS-active substrates by ORC procedures, electrolytes of KCl were generally employed. In contrast, LiClO(4) ones were unsuitable for producing SERS-active substrates. Encouragingly, SERS of Rhodamine 6G (R6G) adsorbed on the roughened Ag substrate prepared in an aqueous solution containing KCl and LiClO(4) electrolytes exhibits a higher intensity by one order of magnitude, as compared with that of R6G adsorbed on a roughened Ag substrate prepared in a solution only containing KCl. Further investigations indicate that the oxidation state of Cl on the roughened Ag substrate demonstrates decided effects on this improved SERS.
Sigma-Aldrich
Lithium perchlorate, SAJ special grade, ≥99.0%
Sigma-Aldrich
Lithium perchlorate, 99.99% trace metals basis
Sigma-Aldrich
Lithium perchlorate, purum p.a., ≥98.0% (calc. based on dry substance, T), powder
Sigma-Aldrich
Lithium perchlorate, ACS reagent, ≥95.0%
Sigma-Aldrich
Lithium perchlorate, battery grade, dry, 99.99% trace metals basis