Merck
Crystallization and preliminary X-ray analysis of Atg3.
Acta crystallographica. Section F, Structural biology and crystallization communications (2006-10-03)
Yuya Yamada, Nobuo N Suzuki, Yuko Fujioka, Yoshinobu Ichimura, Yoshinori Ohsumi, Fuyuhiko Inagaki
ABSTRACT
Atg3 is an E2-like enzyme that catalyzes the conjugation reaction between Atg8 and phosphatidylethanolamine (PE). The Atg8-PE conjugate is essential for autophagy, the bulk degradation process of cytoplasmic components by the vacuolar/lysosomal system. Crystals of Saccharomyces cerevisiae Atg3 have been obtained by the sitting-drop vapour-diffusion method using ammonium sulfate and lithium sulfate as precipitants. A native data set was collected from a single crystal to 2.5 A resolution. The crystals belong to space group P4(1) or P4(3), with unit-cell parameters a = 59.33, c = 115.22 A, and are expected to contain one protein molecule per asymmetric unit.
Sigma-Aldrich
Lithium sulfate, purum p.a., ≥98.0% (T)
Sigma-Aldrich
Lithium sulfate, ≥98.5% (titration)
Sigma-Aldrich
Lithium sulfate, ≥99.99% trace metals basis