Merck
  • High-precision catalysts: regioselective hydroformylation of internal alkenes by encapsulated rhodium complexes.
High-precision catalysts: regioselective hydroformylation of internal alkenes by encapsulated rhodium complexes.
Journal of the American Chemical Society (2006-08-31)
Mark Kuil, Theresa Soltner, Piet W N M van Leeuwen, Joost N H Reek
ABSTRACT
We report the formation of high-precision catalysts using encapsulated rhodium complexes. In the current example, the encapsulated rhodium catalyst shows unprecedented high selectivity in the rhodium-catalyzed hydroformylation of internal alkenes, forming predominantly one of the branched aldehydes. This catalyst system is the first example that is able to discriminate between carbon atoms C3 and C4 in trans-3-octene.
Product Description
Supelco
1-Octene, analytical standard
Sigma-Aldrich
1-Octene, 98%