Merck
  • Low temperature catalytic conversion of methane to methanol by barium sulfate nanotubes supporting sulfates: Pt(SO4)2, HgSO4, Ce(SO4)2 and Pb(SO4)2.
Low temperature catalytic conversion of methane to methanol by barium sulfate nanotubes supporting sulfates: Pt(SO4)2, HgSO4, Ce(SO4)2 and Pb(SO4)2.
Chemical communications (Cambridge, England) (2005-04-28)
Fengbo Li, Guoqing Yuan
ABSTRACT
Barium sulfate nanotubes perform excellently in supporting sulfates (Pt(SO4)2, HgSO4, Ce(SO4)2 and Pb(SO4)2) for low temperature catalytic conversion of methane to methanol under strongly acidic conditions in a conventional gas-phase reactor.
Product Description
Supelco
Mercury(II) sulfate, ACS reagent, ≥99%
Sigma-Aldrich
Mercury(II) sulfate, puriss., ≥98%
Sigma-Aldrich
Lead(II) sulfate, 98%
Sigma-Aldrich
Lead(II) sulfate, 99.995% trace metals basis