Merck
A new assay for lignin-type peroxidases employing the dye azure B.
Applied and environmental microbiology (1992-09-01)
F S Archibald
ABSTRACT
The discovery in 1983 of fungal "ligninases" capable of catalyzing the peroxidation of nonphenolic aromatic lignin components has been seen as a major advance in understanding how certain basidiomycete fungi can completely degrade lignin. The ability of these lignin-type peroxidases to covert millimolar concentrations of veratryl alcohol to veratraldehyde, indicated by a change in the A310 of veratraldehyde, has become the standard assay for routine quantitation of LP activity. A new assay based on the oxidation of micromolar concentrations of the dye Azure B is presented. Although it is as simple and rapid as the veratryl alcohol assay, it appears to overcome some of the shortcomings of that assay. In particular, interference from UV- and short-wavelength visible-light-absorbing materials is greatly reduced and assay specificity is improved.
Sigma-Aldrich
Azure B, prepared by direct synthesis
Sigma-Aldrich
Azure B, certified by the Biological Stain Commission, certified by the BSC