Merck
TS-1 zeolite microengineered reactors for 1-pentene epoxidation.
Chemical communications (Cambridge, England) (2002-07-19)
Yu Shan Susanna Wan, Joseph Lik Hang Chau, Asterios Gavriilidis, King Lun Yeung
ABSTRACT
A zeolite-based microengineered reactor was fabricated and tested for 1-pentene epoxidation over titanium silicalite-1 (TS-1) catalyst, which has been selectively incorporated within the microreactor channel using a new synthesis procedure.
Sigma-Aldrich
1-Pentene, 98%
Supelco
1-Pentene, analytical standard
Sigma-Aldrich
1-Pentene, ≥98.5% (GC)