Merck
  • Propionate reinforces epithelial identity and reduces aggressiveness of lung carcinoma.
Propionate reinforces epithelial identity and reduces aggressiveness of lung carcinoma.
EMBO molecular medicine (2023-09-28)
Vignesh Ramesh, Paradesi Naidu Gollavilli, Luisa Pinna, Mohammad Aarif Siddiqui, Adriana Martinez Turtos, Francesca Napoli, Yasmin Antonelli, Aldo Leal-Egaña, Jesper Foged Havelund, Simon Toftholm Jakobsen, Elisa Le Boiteux, Marco Volante, Nils Joakim Faergeman, Ole N Jensen, Rasmus Siersbaek, Kumar Somyajit, Paolo Ceppi
ABSTRACT
The epithelial-to-mesenchymal transition (EMT) plays a central role in the development of cancer metastasis and resistance to chemotherapy. However, its pharmacological treatment remains challenging. Here, we used an EMT-focused integrative functional genomic approach and identified an inverse association between short-chain fatty acids (propionate and butanoate) and EMT in non-small cell lung cancer (NSCLC) patients. Remarkably, treatment with propionate in vitro reinforced the epithelial transcriptional program promoting cell-to-cell contact and cell adhesion, while reducing the aggressive and chemo-resistant EMT phenotype in lung cancer cell lines. Propionate treatment also decreased the metastatic potential and limited lymph node spread in both nude mice and a genetic NSCLC mouse model. Further analysis revealed that chromatin remodeling through H3K27 acetylation (mediated by p300) is the mechanism underlying the shift toward an epithelial state upon propionate treatment. The results suggest that propionate administration has therapeutic potential in reducing NSCLC aggressiveness and warrants further clinical testing.
Sigma-Aldrich
Anti-PCCB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Troglitazone, ≥98% (HPLC)
Sigma-Aldrich
Histone Acetyl Transferase Inhibitor VII, CTK7A, The Histone Acetyl Transferase Inhibitor VII, CTK7A controls the biological activity of Histone Acetyl Transferase. This small molecule/inhibitor is primarily used for Cell Structure applications.
Sigma-Aldrich
Anti-53BP1 Antibody, clone BP13, clone BP13, Chemicon®, from mouse
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-ZEB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
SAHA, ≥98% (HPLC)