Merck
  • Generation of ventralized human thalamic organoids with thalamic reticular nucleus.
Generation of ventralized human thalamic organoids with thalamic reticular nucleus.
Cell stem cell (2023-04-06)
Ferdi Ridvan Kiral, Bilal Cakir, Yoshiaki Tanaka, Jonghun Kim, Woo Sub Yang, Fabien Wehbe, Young-Jin Kang, Mei Zhong, Gizem Sancer, Sang-Hun Lee, Yangfei Xiang, In-Hyun Park
ABSTRACT
Human brain organoids provide unique platforms for modeling several aspects of human brain development and pathology. However, current brain organoid systems mostly lack the resolution to recapitulate the development of finer brain structures with subregional identity, including functionally distinct nuclei in the thalamus. Here, we report a method for converting human embryonic stem cells (hESCs) into ventral thalamic organoids (vThOs) with transcriptionally diverse nuclei identities. Notably, single-cell RNA sequencing revealed previously unachieved thalamic patterning with a thalamic reticular nucleus (TRN) signature, a GABAergic nucleus located in the ventral thalamus. Using vThOs, we explored the functions of TRN-specific, disease-associated genes patched domain containing 1 (PTCHD1) and receptor tyrosine-protein kinase (ERBB4) during human thalamic development. Perturbations in PTCHD1 or ERBB4 impaired neuronal functions in vThOs, albeit not affecting the overall thalamic lineage development. Together, vThOs present an experimental model for understanding nuclei-specific development and pathology in the thalamus of the human brain.
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 2 Antibody, clone 8G9.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-MAP2 Antibody, clone AP20, clone AP20, Chemicon®, from mouse
Sigma-Aldrich
LDN193189 hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Anti-SK2 (KCa2.2) Antibody, clone K78/29, clone K78/29, from mouse