Merck
  • SAMHD1 controls innate immunity by regulating condensation of immunogenic self RNA.
SAMHD1 controls innate immunity by regulating condensation of immunogenic self RNA.
Molecular cell (2022-09-24)
Shovamayee Maharana, Stefanie Kretschmer, Susan Hunger, Xiao Yan, David Kuster, Sofia Traikov, Thomas Zillinger, Marc Gentzel, Shobha Elangovan, Padmanava Dasgupta, Nagaraja Chappidi, Nadja Lucas, Katharina Isabell Maser, Henrike Maatz, Alexander Rapp, Virginie Marchand, Young-Tae Chang, Yuri Motorin, Norbert Hubner, Gunther Hartmann, Anthony A Hyman, Simon Alberti, Min Ae Lee-Kirsch
ABSTRACT
Recognition of pathogen-derived foreign nucleic acids is central to innate immune defense. This requires discrimination between structurally highly similar self and nonself nucleic acids to avoid aberrant inflammatory responses as in the autoinflammatory disorder Aicardi-Goutières syndrome (AGS). How vast amounts of self RNA are shielded from immune recognition to prevent autoinflammation is not fully understood. Here, we show that human SAM-domain- and HD-domain-containing protein 1 (SAMHD1), one of the AGS-causing genes, functions as a single-stranded RNA (ssRNA) 3'exonuclease, the lack of which causes cellular RNA accumulation. Increased ssRNA in cells leads to dissolution of RNA-protein condensates, which sequester immunogenic double-stranded RNA (dsRNA). Release of sequestered dsRNA from condensates triggers activation of antiviral type I interferon via retinoic-acid-inducible gene I-like receptors. Our results establish SAMHD1 as a key regulator of cellular RNA homeostasis and demonstrate that buffering of immunogenic self RNA by condensates regulates innate immune responses.
Product Description
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Sigma-Aldrich
Anti-SAMHD1 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
ISRIB, ≥98% (HPLC)