Merck
  • Direct analysis of phthalate esters in vegetable oils by means of comprehensive two-dimensional gas chromatography combined with triple quadrupole mass spectrometry.
Direct analysis of phthalate esters in vegetable oils by means of comprehensive two-dimensional gas chromatography combined with triple quadrupole mass spectrometry.
Food chemistry (2022-07-24)
Alessia Arena, Mariosimone Zoccali, Luigi Mondello, Peter Q Tranchida
ABSTRACT
Phthalic acid esters (PAEs), known as phthalates, have a variety of industrial applications, mainly related to their ability to increase plastic softness,flexibility, and durability. With regard to toxicological aspects, several studies suggest that phthalates would act as endocrine-disrupting chemicals, and they have been correlated to several forms of cancer. Since the analysis of PAEs in food is challenging, the aim of this study was the development of a simple and direct method, with no sample preparation (only dilution withsolvent), for the analysis of targeted phthalates in vegetable oils, by using cryogenic-modulation comprehensive two-dimensional gas chromatography combined with triple quadrupole mass spectrometry. The method was characterized by good repeatability values, limit of quantification values ranging between 0.06 and 2.10 mg/kg, accuracy values ranging from -9.2 to 10.4%, and a matrix effectranging from -4 to 78%.The developed technique was used for the analysis of 27 vegetable oils.
Product Description
Supelco
SLB®-5ms Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.10 μm
Supelco
3,5-Di-tert-butyl-4-hydroxytoluene, analytical standard
Supelco
Diisodecyl phthalate, for ion-selective electrodes, Selectophore, ≥99.0%
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Supelco
Benzyl butyl phthalate, analytical standard