Merck
  • Caveolin-1 is involved in fatty infiltration and bone-tendon healing of rotator cuff tear.
Caveolin-1 is involved in fatty infiltration and bone-tendon healing of rotator cuff tear.
Molecular medicine (Cambridge, Mass.) (2023-03-16)
Shanhong Fang, Mengqiang You, Jie Wei, Peng Chen
ABSTRACT
Caveolin-1 has been predicted, based on RNA transcriptome sequencing, as a key gene in rotator cuff tear (RCT) and it is related to fatty infiltration. This study aims to elucidate the upstream and downstream mechanism of Caveolin-1 in fatty infiltration and bone-tendon healing after RCT in rat models. Differentially expressed genes related to RCT were screened, followed by functional enrichment analysis and protein-protein interaction analysis. GATA6 was overexpressed and Caveolin-1 was knocked down in tendon stem cells (TSCs) to evaluate their effects on the adipogenic differentiation of TSCs. In addition, a RCT rat model was constructed and injected with lentivirus carrying oe-GATA6, oe-Caveolin-1 alone or in combination to assess their roles in fatty infiltration and bone-tendon healing. RESULTS AND CONCLUSION: Caveolin-1 was identified as a key gene involved in the RCT process. In vitro results demonstrated that Caveolin-1 knockdown inhibited adipogenic differentiation of TSCs by activating the cAMP/PKA pathway. GATA6 inhibited the transcription of Caveolin-1 and inhibited its expression, thus suppressing the adipogenic differentiation of TSCs. In vivo data confirmed that GATA6 overexpression activated the cAMP/PKA pathway by downregulating Caveolin-1 and consequently repressed fatty infiltration, promoted bone-tendon healing, improved biomechanical properties and reduced the rupture risk of injured tendon in rats after RCT. Overall, this study provides novel insights into the mechanistic action of Caveolin-1 in the fatty infiltration and bone-tendon healing after RCT.
Sigma-Aldrich
H-89 dihydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Collagenase type I, The collagenase type I (from Clostridium histolyticum) is a crude collagenase preparation that can be used for the isolation of primary cells or for tissue dissociation by enzymatic means.
Sigma-Aldrich
Anti-HSL antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-PKA CAT (pThr197) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-HSL (pSer552) antibody produced in rabbit, affinity isolated antibody