Merck
  • Computer-aided engineering of staphylokinase toward enhanced affinity and selectivity for plasmin.
Computer-aided engineering of staphylokinase toward enhanced affinity and selectivity for plasmin.
Computational and structural biotechnology journal (2022-04-08)
Dmitri Nikitin, Jan Mican, Martin Toul, David Bednar, Michaela Peskova, Patricia Kittova, Sandra Thalerova, Jan Vitecek, Jiri Damborsky, Robert Mikulik, Sarel J Fleishman, Zbynek Prokop, Martin Marek
ABSTRACT
Cardio- and cerebrovascular diseases are leading causes of death and disability, resulting in one of the highest socio-economic burdens of any disease type. The discovery of bacterial and human plasminogen activators and their use as thrombolytic drugs have revolutionized treatment of these pathologies. Fibrin-specific agents have an advantage over non-specific factors because of lower rates of deleterious side effects. Specifically, staphylokinase (SAK) is a pharmacologically attractive indirect plasminogen activator protein of bacterial origin that forms stoichiometric noncovalent complexes with plasmin, promoting the conversion of plasminogen into plasmin. Here we report a computer-assisted re-design of the molecular surface of SAK to increase its affinity for plasmin. A set of computationally designed SAK mutants was produced recombinantly and biochemically characterized. Screening revealed a pharmacologically interesting SAK mutant with ∼7-fold enhanced affinity toward plasmin, ∼10-fold improved plasmin selectivity and moderately higher plasmin-generating efficiency in vitro. Collectively, the results obtained provide a framework for SAK engineering using computational affinity-design that could pave the way to next-generation of effective, highly selective, and less toxic thrombolytics.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
α-Lactalbumin from bovine milk, Type I, ≥85% (PAGE), lyophilized powder