Merck
  • CD36-mediated ferroptosis dampens intratumoral CD8+ T cell effector function and impairs their antitumor ability.
CD36-mediated ferroptosis dampens intratumoral CD8+ T cell effector function and impairs their antitumor ability.
Cell metabolism (2021-03-11)
Xingzhe Ma, Liuling Xiao, Lintao Liu, Lingqun Ye, Pan Su, Enguang Bi, Qiang Wang, Maojie Yang, Jianfei Qian, Qing Yi
ABSTRACT
Understanding the mechanisms underlying how T cells become dysfunctional in a tumor microenvironment (TME) will greatly benefit cancer immunotherapy. We found that increased CD36 expression in tumor-infiltrating CD8+ T cells, which was induced by TME cholesterol, was associated with tumor progression and poor survival in human and murine cancers. Genetic ablation of Cd36 in effector CD8+ T cells exhibited increased cytotoxic cytokine production and enhanced tumor eradication. CD36 mediated uptake of fatty acids by tumor-infiltrating CD8+ T cells in TME, induced lipid peroxidation and ferroptosis, and led to reduced cytotoxic cytokine production and impaired antitumor ability. Blocking CD36 or inhibiting ferroptosis in CD8+ T cells effectively restored their antitumor activity and, more importantly, possessed greater antitumor efficacy in combination with anti-PD-1 antibodies. This study reveals a new mechanism of CD36 regulating the function of CD8+ effector T cells and therapeutic potential of targeting CD36 or inhibiting ferroptosis to restore T cell function.
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
1S,3R-RSL 3, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
FIN56, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Fatty Acid Uptake Kit, sufficient for 100 fluorometric tests
Pricing and availability is not currently available.
Sigma-Aldrich
Ferrostatin-1, ≥95% (HPLC)
Pricing and availability is not currently available.