Merck
  • MUC4 is overexpressed in idiopathic pulmonary fibrosis and collaborates with transforming growth factor β inducing fibrotic responses.
MUC4 is overexpressed in idiopathic pulmonary fibrosis and collaborates with transforming growth factor β inducing fibrotic responses.
Mucosal immunology (2020-09-06)
Javier Milara, Beatriz Ballester, M J Safont, Enrique Artigues, Juan Escrivá, Esteban Morcillo, Julio Cortijo
ABSTRACT
Several mucins are implicated in idiopathic pulmonary fibrosis (IPF); however, there is no evidence regarding the role of MUC4 in the development of IPF. Here we demonstrated that MUC4 was overexpressed in IPF patients (n = 22) compared with healthy subjects (n = 21) and located in pulmonary arteries, bronchial epithelial cells, fibroblasts, and hyperplastic alveolar type II cells. Decreased expression of MUC4 using siRNA-MUC4 inhibited the mesenchymal/myofibroblast transformations of alveolar type II A549 cells and lung fibroblasts, as well as cell senescence and fibroblast proliferation induced by TGF-β1. The induction of the overexpression of MUC4 increased the effects of TGF-β1 on mesenchymal/myofibroblast transformations and cell senescence. MUC4 overexpression and siRNA-MUC4 gene silencing increased or decreased, respectively, the phosphorylation of TGFβRI and SMAD3, contributing to smad-binding element activation. Immunoprecipitation analysis and confocal immunofluorescence showed the formation of a protein complex between MUC4β/p-TGFβRI and p-SMAD3 in the cell membrane after TGF-β1 stimulation and in lung tissue from IPF patients. Bleomycin-induced lung fibrosis was reduced in mice transiently transfected with siRNA-MUC4. This study shows that MUC4 expression is enhanced in IPF and promotes fibrotic processes in collaboration with TGF-β1 canonical pathway that could be an attractive druggable target for human IPF.
Sigma-Aldrich
Transforming Growth Factor-β1 human, TGF-β1, recombinant, expressed in CHO cells, powder, suitable for cell culture
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Senescence Cells Histochemical Staining Kit, sufficient for 100 tests
Sigma-Aldrich
PhosphoDetect Anti-Smad3 (pSer423/425) Rabbit mAb (C25A9), liquid, clone C25A9, Calbiochem®
Sigma-Aldrich
Anti-Collagen Type I Rabbit pAb, liquid, Calbiochem®