Merck
  • Hippocampal interneurons co-express transcripts encoding the alpha7 nicotinic receptor subunit and the cannabinoid receptor 1.
Hippocampal interneurons co-express transcripts encoding the alpha7 nicotinic receptor subunit and the cannabinoid receptor 1.
Neuroscience (2008-01-29)
M Morales, K Hein, Z Vogel
ABSTRACT
The notion of functional interactions between the alpha7 nicotinic acetylcholine (alpha7 nACh) and the cannabinoid systems is emerging from recent in vitro and in vivo studies. Both the alpha7 nACh receptor and the cannabinoid receptor 1 (CB1) are highly expressed in the hippocampus. To begin addressing possible anatomical interactions between the alpha7 nACh and the cannabinoid systems in the rat hippocampus, we investigated the distribution of neurons expressing alpha7 nACh mRNA in relation to those containing CB1 mRNA. By in situ hybridization we found that the alpha7 nACh mRNA is diffusely expressed in principal neurons and is highly expressed in a subset of interneurons. We observed that the pattern of distribution of hippocampal interneurons co-expressing transcripts encoding alpha7 nACh and glutamate decarboxylase (GAD; synthesizing enzyme of GABA) closely resembles the one displayed by interneurons expressing CB1 mRNA. By double in situ hybridization we established that the majority of hippocampal interneurons expressing alpha7 nACh mRNA have high levels of CB1 mRNA. As CB1 interneurons contain cholecystokinin (CCK), we investigated the degree of cellular co-expression of alpha7 nACh mRNA and CCK, and found that the cellular co-existence of alpha7 nACh and CCK varies within the different layers of the hippocampus. In summary, we established that most of the hippocampal alpha7 nACh expressing interneurons are endowed with CB1 mRNA. We found that these alpha7 nACh/CB1 interneurons are the major subpopulation of hippocampal interneurons expressing CB1 mRNA. The alpha7 nACh expressing interneurons represent half of the detected population of CCK containing neurons in the hippocampus. Since it is well established that the vast majority of hippocampal interneurons expressing CB1 mRNA have 5-HT type 3 (5-HT3) receptors, we conclude that these hippocampal alpha7 nACh/5HT3/CB1/CCK interneurons correspond to those previously postulated to relay inputs from diverse cortical and subcortical regions about emotional, motivational, and physiological states.
Sigma-Aldrich
Albumin from rat serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from porcine serum, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from rabbit serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from rat serum, lyophilized powder, essentially fatty acid free, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from rat serum, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from mouse serum, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from mouse serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from porcine serum, lyophilized powder, 96-99% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from porcine serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from rabbit serum, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)