Merck
  • Upregulation of Antioxidant Capacity and Nucleotide Precursor Availability Suffices for Oncogenic Transformation.
Upregulation of Antioxidant Capacity and Nucleotide Precursor Availability Suffices for Oncogenic Transformation.
Cell metabolism (2020-11-08)
Yang Zhang, Yi Xu, Wenyun Lu, Jonathan M Ghergurovich, Lili Guo, Ian A Blair, Joshua D Rabinowitz, Xiaolu Yang
ABSTRACT
The emergence of cancer from diverse normal tissues has long been rationalized to represent a common set of fundamental processes. However, these processes are not fully defined. Here, we show that forced expression of glucose-6-phosphate dehydrogenase (G6PD) affords immortalized mouse and human cells anchorage-independent growth in vitro and tumorigenicity in animals. Mechanistically, G6PD augments the NADPH pool by stimulating NAD+ kinase-mediated NADP+ biosynthesis in addition to converting NADP+ to NADPH, bolstering antioxidant defense. G6PD also increases nucleotide precursor levels through the production of ribose and NADPH, promoting cell proliferation. Supplementation of antioxidants or nucleosides suffices to convert immortalized mouse and human cells into a tumorigenic state, and supplementation of both is required when their overlapping metabolic consequences are minimized. These results suggest that normal cells have a limited capacity for redox balance and nucleotide synthesis, and overcoming this limit might represent a key aspect of oncogenic transformation.
Sigma-Aldrich
Cell Counting Kit - 8, for quantitation of viable cell number in proliferation and cytotoxicity assays
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
NAD Kinase human, recombinant, expressed in E. coli, His tagged, >95% (SDS-PAGE)
Sigma-Aldrich
G6PD human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture