Merck
  • Food Reward Chemistry Explains a Novel Pollinator Shift and Vestigialization of Long Floral Spurs in an Orchid.
Food Reward Chemistry Explains a Novel Pollinator Shift and Vestigialization of Long Floral Spurs in an Orchid.
Current biology : CB (2020-11-07)
Miguel Castañeda-Zárate, Steven D Johnson, Timotheüs van der Niet
ABSTRACT
During the evolutionary history of flowering plants, transitions between pollinator groups (pollinator shifts) have been frequent,1 and contributed to the spectacular radiation of angiosperms.2 Although the evolution of floral traits during pollinator shifts has been studied in real time under controlled laboratory conditions,3 it is challenging to study in nature and therefore poorly understood.4-7 Using a comparative, multidisciplinary approach, we dissect the evolution of floral traits during a pollinator shift in the long-spurred African orchid Satyrium longicauda. Phylogenetic analysis and ecological experiments revealed a shift from moth- to oil-collecting bee pollination. Remarkably, flowers of the bee-pollinated form are similar in morphology, color, and overall volatile chemistry to those of moth-pollinated forms, but differ in having spurs that are mostly devoid of nectar, and have an elevated presence of the oil-derived compound diacetin, which oil-collecting bees use as a cue for oil presence.8 Experiments demonstrated that long spurs are critical for pollination of a moth-pollinated form, but are not needed for pollination of the bee-pollinated form. We conclude that the pollinator shift in Satyrium was mediated by a switch in chemistry of the pollinator reward. The ancestral presence of diacetin might have served as a pre-adaptation for bee pollination, whereas the current mismatch between flower morphology and bees is due to the retention of vestigial floral spurs. These results elucidate the sequence of floral evolution in the early stages of pollinator shifts and help to explain the assembly of suites of co-varying traits through pre-adaptation and vestigialization.9-12.
Sigma-Aldrich
Diacetin
Sigma-Aldrich
Glyceryl triacetate, ≥99%
Sigma-Aldrich
Glyceryl triacetate, ≥99.0% (GC)
Supelco
C7 - C40 Saturated Alkanes Standard, certified reference material, 1000 μg/mL each component in hexane
Sigma-Aldrich
Diacetin, technical grade, 50%