Merck
  • New Electroactive Polymers with Electronically Isolated 4,7-Diarylfluorene Chromophores as Positive Charge Transporting Layer Materials for OLEDs.
New Electroactive Polymers with Electronically Isolated 4,7-Diarylfluorene Chromophores as Positive Charge Transporting Layer Materials for OLEDs.
Molecules (Basel, Switzerland) (2021-04-04)
Gintare Krucaite, Daiva Tavgeniene, Dovydas Blazevicius, Baohua Zhang, Aivars Vembris, Saulius Grigalevicius
ABSTRACT
A group of polyethers containing electroactive pendent 4,7-diarylfluorene chromophores have been prepared by the multi-step synthetic route. Full characterization of their structures has been presented. The polymeric materials represent derivatives of high thermal stability with initial thermal degradation temperatures in a range of 392-397 °C. Glass transition temperatures of the amorphous polymers range from 28 °C to 63 °C and depend on structures of the 4,7-diarylfluorene chromophores. Electron photoemission spectra of thin layers of the electroactive derivatives showed ionization potentials in the range of 5.8-6.0 eV. Hole injecting/transporting properties of the prepared polymeric materials were confirmed during formation of organic light-emitting diodes with tris(quinolin-8-olato)aluminium (Alq3) as a green emitter, which also serves as an electron transporting layer. The device using hole-transporting polymer with electronically isolated 2,7-di(4-biphenyl)fluorene chromophores demonstrated the best overall performance with low turn on voltage of 3 V, high current efficiency exceeding 1.7 cd/A, and with maximum brightness over 200 cd/m2. The organic light-emitting diode (OLED) characteristics were measured in non-optimized test devices. The efficiencies could be further improved by an optimization of device structure, formation conditions, and encapsulation of the devices.
Sigma-Aldrich
4-Biphenylboronic acid, ≥95.0%
Sigma-Aldrich
3-Methyl-3-oxetanemethanol, 98%
Sigma-Aldrich
2,7-Dibromofluorene, 97%