Merck
  • Combretastatin-like chalcones as inhibitors of microtubule polymerization. Part 1: synthesis and biological evaluation of antivascular activity.
Combretastatin-like chalcones as inhibitors of microtubule polymerization. Part 1: synthesis and biological evaluation of antivascular activity.
Bioorganic & medicinal chemistry (2009-10-20)
Sylvie Ducki, David Rennison, Meiko Woo, Alexander Kendall, Jérémie Fournier Dit Chabert, Alan T McGown, Nicholas J Lawrence
ABSTRACT
The alpha-methyl chalcone SD400 is a potent inhibitor of tubulin assembly and possesses potent anticancer activity. Various chalcone analogues were synthesized and evaluated for their cell growth inhibitory properties against the K562 human chronic myelogenous leukemia cell line (SD400, IC(50) 0.21nM; combretastatin A4 CA4, IC(50) 2.0nM). Cell cycle analysis by flow cytometry indicated that these agents are antimitotic (SD400, 83% of the cells are in G(2)/M phase; CA4 90%). They inhibit tubulin assembly at low concentration (SD400, IC(50) 0.46microM; CA4, 0.10microM) and compete with [(3)H]colchicine for binding to tubulin (8% [(3)H]colchicine remained bound to tubulin after competition with SD400 or CA4). Upon treatment with SD400, remarkable cell shape changes were elicited in HUVEC cells, consistent with vasculature damaging activity.
Sigma-Aldrich
trans-Chalcone, 97%