Merck
  • New uracil dimers showing erythroid differentiation inducing activities.
New uracil dimers showing erythroid differentiation inducing activities.
Journal of medicinal chemistry (2008-12-17)
Alessandro Accetta, Roberto Corradini, Stefano Sforza, Tullia Tedeschi, Eleonora Brognara, Monica Borgatti, Roberto Gambari, Rosangela Marchelli
ABSTRACT
The synthesis of C5 linked uracil dimers was carried out according to a model developed in order to bind adenine in DNA. N1-Alkylated uracil derivatives were synthesized from isoorotic acid (uracil-5-carboxylic acid) or thymine. The carboxylic acid derivatives were condensed with diamines in order to produce dimeric compounds or with monoamines in order to obtain reference monomeric compounds. Some of the derivatives, in particular the uracil dimers, showed antiproliferative and erythroid differentiation induction properties towards human chronic myelogenous leukemia K562 cells, thus indicating that these compounds could represent a new class of drugs useful for the development of antitumor therapy based on the ability to induce terminal differentiation.
Product Description
Supelco
Butyric acid, analytical standard
Sigma-Aldrich
Butyric acid, ≥99%, FG
Sigma-Aldrich
Butyric acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
Butyric acid, ≥99%
Sigma-Aldrich
2,4-Dihydroxypyrimidine-5-carboxylic acid, 95%
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)