Merck
Room temperature 3D printing of super-soft and solvent-free elastomers.
Science advances (2020-11-15)
Renxuan Xie, Sanjoy Mukherjee, Adam E Levi, Veronica G Reynolds, Hengbin Wang, Michael L Chabinyc, Christopher M Bates
ABSTRACT
Super-soft elastomers derived from bottlebrush polymers show promise as advanced materials for biomimetic tissue and device applications, but current processing strategies are restricted to simple molding. Here, we introduce a design concept that enables the three-dimensional (3D) printing of super-soft and solvent-free bottlebrush elastomers at room temperature. The key advance is a class of inks comprising statistical bottlebrush polymers that self-assemble into well-ordered body-centered cubic sphere phases. These soft solids undergo sharp and reversible yielding at 20°C in response to shear with a yield stress that can be tuned by manipulating the length scale of microphase separation. The addition of a soluble photocrosslinker allows complete ultraviolet curing after extrusion to form super-soft elastomers with near-perfect recoverable elasticity well beyond the yield strain. These structure-property design rules create exciting opportunities to tailor the performance of 3D-printed elastomers in ways that are not possible with current materials and processes.
Sigma-Aldrich
Succinic anhydride, ≥99% (GC)
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Sigma-Aldrich
Poly(dimethylsiloxane), bis(hydroxyalkyl) terminated, average Mn ~5,600
Sigma-Aldrich
4-Benzoylbenzoic acid, 99%