Merck
  • Contributions of the combined effects of topoisomerase mutations toward fluoroquinolone resistance in Escherichia coli.
Contributions of the combined effects of topoisomerase mutations toward fluoroquinolone resistance in Escherichia coli.
Antimicrobial agents and chemotherapy (2007-08-08)
Sonia K Morgan-Linnell, Lynn Zechiedrich
ABSTRACT
In defined, isogenic strains, at least three mutations, two of which must be in gyrA, were required to exceed the CLSI breakpoint for fluoroquinolone resistance. Strains with double mutations in both gyrA and parC had even higher MICs of fluoroquinolones than strains with totals of three mutations.
Product Description
Supelco
Norfloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Sigma-Aldrich
Sparfloxacin, ≥98.0% (HPLC)
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)