Merck
Reproducibility of MDM2 and CDK4 staining in soft tissue tumors.
American journal of clinical pathology (2006-05-19)
Matthieu Bui Nguyen Binh, Xavier Sastre Garau, Louis Guillou, Alain Aurias, Jean-Michel Coindre
ABSTRACT
MDM2 and CDK4 immunostaining can be useful adjuncts in diagnosing liposarcoma among soft tissue neoplasms. We examined the reproducibility of MDM2 and CDK4 staining between 2 laboratories and between tissue microarrays and whole tissue sections. Sixty-two soft tissue tumors were immunostained at the Bergonié Institute, Bordeaux, France, and the Curie Institute, Paris, France. We also examined 203 soft tissue neoplasms on standard tissue sections and tissue microarrays. There was high concordance of results obtained from the 2 laboratories (with 2 different pathologists) for MDM2 (kappa, 0.93) and CDK4 (kappa, 0.8) staining. There also was excellent concordance between results on tissue microarray and on whole tissue sections for MDM2 (kappa, 0.80) and CDK4 (kappa, 0.93). Immunostaining for MDM2 and CDK4 is a reproducible technique that may be exported to different laboratories for routine use. Tissue microarray is indicated for studying large series.
Sigma-Aldrich
MDM2 (IF2) Mouse Monoclonal Antibody