Merck
  • Inhibition of bacterial adhesion and biofilm formation by a textured fluorinated alkoxyphosphazene surface.
Inhibition of bacterial adhesion and biofilm formation by a textured fluorinated alkoxyphosphazene surface.
Bioactive materials (2020-10-01)
Meixian Tang, Chen Chen, Jieru Zhu, Harry R Allcock, Christopher A Siedlecki, Li-Chong Xu
ABSTRACT
The utilization of biomaterials in implanted blood-contacting medical devices often induces a persistent problem of microbial infection, which results from bacterial adhesion and biofilm formation on the surface of biomaterials. In this research, we developed new fluorinated alkoxyphosphazene materials, specifically poly[bis(octafluoropentoxy) phosphazene] (OFP) and crosslinkable OFP (X-OFP), with improved mechanical properties, and further modified the surface topography with ordered pillars to improve the antibacterial properties. Three X-OFP materials, X-OFP3.3, X-OFP8.1, X-OFP13.6, with different crosslinking densities were synthesized, and textured films with patterns of 500/500/600 nm (diameter/spacing/height) were fabricated via a two stage soft lithography molding process. Experiments with 3 bacterial strains: Staphylococcal epidermidis, Staphylococcal aureus, and Pseudomonas aeruginosa showed that bacterial adhesion coefficients were significantly lower on OFP and X-OFP smooth surfaces than on the polyurethane biomaterial, and surface texturing further reduced bacterial adhesion due to the reduction in accessible surface contact area. Furthermore the anti-bacterial adhesion effect shows a positive relationship with the crosslinking degree. Biofilm formation on the substrates was examined using a CDC biofilm reactor for 7 days and no biofilm formation was observed on textured X-OFP biomaterials. The results suggested that the combination of fluorocarbon chemistry and submicron topography modification in textured X-OFP materials may provide a practical approach to improve the biocompatibility of current biomaterials with significant reduction in risk of pathogenic infection.
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Sigma-Aldrich
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol, 98%
Sigma-Aldrich
2-Allylphenol, 98%