Merck
  • Role of inner solvation sheath within salt-solvent complexes in tailoring electrode/electrolyte interphases for lithium metal batteries.
Role of inner solvation sheath within salt-solvent complexes in tailoring electrode/electrolyte interphases for lithium metal batteries.
Proceedings of the National Academy of Sciences of the United States of America (2020-11-05)
Xiaodi Ren, Peiyuan Gao, Lianfeng Zou, Shuhong Jiao, Xia Cao, Xianhui Zhang, Hao Jia, Mark H Engelhard, Bethany E Matthews, Haiping Wu, Hongkyung Lee, Chaojiang Niu, Chongmin Wang, Bruce W Arey, Jie Xiao, Jun Liu, Ji-Guang Zhang, Wu Xu
ABSTRACT
Functional electrolyte is the key to stabilize the highly reductive lithium (Li) metal anode and the high-voltage cathode for long-life, high-energy-density rechargeable Li metal batteries (LMBs). However, fundamental mechanisms on the interactions between reactive electrodes and electrolytes are still not well understood. Recently localized high-concentration electrolytes (LHCEs) are emerging as a promising electrolyte design strategy for LMBs. Here, we use LHCEs as an ideal platform to investigate the fundamental correlation between the reactive characteristics of the inner solvation sheath on electrode surfaces due to their unique solvation structures. The effects of a series of LHCEs with model electrolyte solvents (carbonate, sulfone, phosphate, and ether) on the stability of high-voltage LMBs are systematically studied. The stabilities of electrodes in different LHCEs indicate the intrinsic synergistic effects between the salt and the solvent when they coexist on electrode surfaces. Experimental and theoretical analyses reveal an intriguing general rule that the strong interactions between the salt and the solvent in the inner solvation sheath promote their intermolecular proton/charge transfer reactions, which dictates the properties of the electrode/electrolyte interphases and thus the battery performances.
Sigma-Aldrich
Triethyl phosphate, ReagentPlus®, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Pricing and availability is not currently available.