Merck
  • Neurotoxic microglia promote TDP-43 proteinopathy in progranulin deficiency.
Neurotoxic microglia promote TDP-43 proteinopathy in progranulin deficiency.
Nature (2020-09-01)
Jiasheng Zhang, Dmitry Velmeshev, Kei Hashimoto, Yu-Hsin Huang, Jeffrey W Hofmann, Xiaoyu Shi, Jiapei Chen, Andrew M Leidal, Julian G Dishart, Michelle K Cahill, Kevin W Kelley, Shane A Liddelow, William W Seeley, Bruce L Miller, Tobias C Walther, Robert V Farese, J Paul Taylor, Erik M Ullian, Bo Huang, Jayanta Debnath, Torsten Wittmann, Arnold R Kriegstein, Eric J Huang
ABSTRACT
Aberrant aggregation of the RNA-binding protein TDP-43 in neurons is a hallmark of frontotemporal lobar degeneration caused by haploinsufficiency in the gene encoding progranulin1,2. However, the mechanism leading to TDP-43 proteinopathy remains unclear. Here we use single-nucleus RNA sequencing to show that progranulin deficiency promotes microglial transition from a homeostatic to a disease-specific state that causes endolysosomal dysfunction and neurodegeneration in mice. These defects persist even when Grn-/- microglia are cultured ex vivo. In addition, single-nucleus RNA sequencing reveals selective loss of excitatory neurons at disease end-stage, which is characterized by prominent nuclear and cytoplasmic TDP-43 granules and nuclear pore defects. Remarkably, conditioned media from Grn-/- microglia are sufficient to promote TDP-43 granule formation, nuclear pore defects and cell death in excitatory neurons via the complement activation pathway. Consistent with these results, deletion of the genes encoding C1qa and C3 mitigates microglial toxicity and rescues TDP-43 proteinopathy and neurodegeneration. These results uncover previously unappreciated contributions of chronic microglial toxicity to TDP-43 proteinopathy during neurodegeneration.
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 30,000-70,000
Sigma-Aldrich
Anti-Tom20/Tomm20 Antibody, clone 2F8.1, clone 2F8.1, from mouse
Sigma-Aldrich
Anti-Complement C3b/iC3b Antibody, clone 3E7, neutralizing, clone 3E7, from mouse
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Goat Anti-Mouse IgG, H&L Chain Specific Peroxidase Conjugate, liquid, Calbiochem®
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse