Merck
  • The Presence of Survivin on B Cells from Myasthenia Gravis Patients and the Potential of an Antibody to a Modified Survivin Peptide to Alleviate Weakness in an Animal Model.
The Presence of Survivin on B Cells from Myasthenia Gravis Patients and the Potential of an Antibody to a Modified Survivin Peptide to Alleviate Weakness in an Animal Model.
Journal of immunology (Baltimore, Md. : 1950) (2020-08-26)
Xiangyang Zhang, Michael Ciesielski, Robert A Fenstermaker, Henry J Kaminski, Linda L Kusner
ABSTRACT
Myasthenia gravis (MG) is an autoimmune disease in which Abs target neuromuscular junction proteins, in particular the acetylcholine receptor. We previously identified the antiapoptotic protein survivin in the autoreactive B cells and plasma cells of MG patients. To further define the role of survivin in MG, we have assessed PBMCs from 29 patients with MG and 15 controls. We confirmed the increased expression of survivin in CD20+ lymphocytes from MG patients compared with controls. Furthermore, the CD20+ population of cells from MG patients contained a higher percentage of extracellular survivin compared with controls. The analysis of CD4+ cells showed an increased percentage of intracellular survivin in MG patients compared with controls, whereas the extracellular survivin CD4+ percentage was unaffected. In an experimental mouse model of MG, we assessed the therapeutic potential of an Ab raised to a modified survivin peptide but cross-reactive to survivin. Ab treatment reduced disease severity, lowered acetylcholine receptor-specific Abs, and decreased CD19+ survivin+ splenocytes. The ability to target survivin through Ab recognition of autoreactive cells offers the potential for a highly specific therapeutic agent for MG.
Product Description
Corning® 96 Well EIA/RIA Assay Microplate, flat bottom clear, polystyrene, high binding surface, pkg of (individually wrapped), non-sterile, lid: no
Sigma-Aldrich
Phosphate buffered saline, BioPerformance Certified, pH 7.4
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Freund′s Adjuvant, Complete, cell suspension
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)