Merck
Removal Ability and Resistance to Cinnamic and Vanillic Acids by Fungi.
Microorganisms (2020-06-25)
Barbara Speranza, Francesca Cibelli, Antonietta Baiano, Antonia Carlucci, Maria Luisa Raimondo, Daniela Campaniello, Ilaria Viggiani, Antonio Bevilacqua, Maria Rosaria Corbo
ABSTRACT
Twelve fungal strains were assayed to investigate their resistance to cinnamic and vanillic acids and their ability to remove these compounds from a liquid medium. In a first step, the effect of the two aromatic acids (1 g/L) on the fungal growth kinetic was studied. The results were modelled through a logistic like function (Dantigny equation) to estimate τ, which is the time to the half-maximum colony diameter. The key findings of this part were as follows: (i) generally, cinnamic acid exerted a stronger effect than vanillic acid; (ii) aromatic acids exerted a delay on the growth of some fungi and only one strain (Athelia rolfsii) was completely inhibited. In the second part, fungi were assayed to investigate their ability to remove cinnamic and vanillic acids (ca. 350 mg/kg) from liquid media at pH 3.5. The results indicated that the most efficient fungi were Aspergillus niger and Lasiodiplodia theobromae.
Sigma-Aldrich
trans-Cinnamic acid, 97%
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)