Merck
  • Chronic activation of endothelial MAPK disrupts hematopoiesis via NFKB dependent inflammatory stress reversible by SCGF.
Chronic activation of endothelial MAPK disrupts hematopoiesis via NFKB dependent inflammatory stress reversible by SCGF.
Nature communications (2020-02-06)
Pradeep Ramalingam, Michael G Poulos, Elisa Lazzari, Michael C Gutkin, David Lopez, Christopher C Kloss, Michael J Crowley, Lizabeth Katsnelson, Ana G Freire, Matthew B Greenblatt, Christopher Y Park, Jason M Butler
ABSTRACT
Inflammatory signals arising from the microenvironment have emerged as critical regulators of hematopoietic stem cell (HSC) function during diverse processes including embryonic development, infectious diseases, and myelosuppressive injuries caused by irradiation and chemotherapy. However, the contributions of cellular subsets within the microenvironment that elicit niche-driven inflammation remain poorly understood. Here, we identify endothelial cells as a crucial component in driving bone marrow (BM) inflammation and HSC dysfunction observed following myelosuppression. We demonstrate that sustained activation of endothelial MAPK causes NF-κB-dependent inflammatory stress response within the BM, leading to significant HSC dysfunction including loss of engraftment ability and a myeloid-biased output. These phenotypes are resolved upon inhibition of endothelial NF-κB signaling. We identify SCGF as a niche-derived factor that suppresses BM inflammation and enhances hematopoietic recovery following myelosuppression. Our findings demonstrate that chronic endothelial inflammation adversely impacts niche activity and HSC function which is reversible upon suppression of inflammation.
Sigma-Aldrich
Pimonidazole, ≥98% (HPLC)
Sigma-Aldrich
Sunflower seed oil from Helianthus annuus
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Biotin, ≥99% (HPLC), lyophilized powder
Sigma-Aldrich
Tamoxifen, ≥99%
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Corning® 1 x 8 Stripwell 96 well plates, high binding surface, clear
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
Evans Blue, Dye content ≥75 %