Merck
  • Kindlin-2 Regulates the Growth of Breast Cancer Tumors by Activating CSF-1-Mediated Macrophage Infiltration.
Kindlin-2 Regulates the Growth of Breast Cancer Tumors by Activating CSF-1-Mediated Macrophage Infiltration.
Cancer research (2017-07-09)
Khalid Sossey-Alaoui, Elzbieta Pluskota, Katarzyna Bialkowska, Dorota Szpak, Yvonne Parker, Chevaun D Morrison, Daniel J Lindner, William P Schiemann, Edward F Plow
ABSTRACT
Interplay between tumor cells and host cells in the tumor microenvironment dictates the development of all cancers. In breast cancer, malignant cells educate host macrophages to adopt a protumorigenic phenotype. In this study, we show how the integrin-regulatory protein kindlin-2 (FERMT2) promotes metastatic progression of breast cancer through the recruitment and subversion of host macrophages. Kindlin-2 expression was elevated in breast cancer biopsy tissues where its levels correlated with reduced patient survival. On the basis of these observations, we used CRISPR/Cas9 technology to ablate Kindlin-2 expression in human MDA-MB-231 and murine 4T1 breast cancer cells. Kindlin-2 deficiency inhibited invasive and migratory properties in vitro without affecting proliferation rates. However, in vivo tumor outgrowth was inhibited by >80% in a manner associated with reduced macrophage infiltration and secretion of the macrophage attractant and growth factor colony-stimulating factor-1 (CSF-1). The observed loss of CSF-1 appeared to be caused by a more proximal deficiency in TGFĪ²-dependent signaling in Kindlin-2-deficient cells. Collectively, our results illuminate a Kindlin-2/TGFĪ²/CSF-1 signaling axis employed by breast cancer cells to capture host macrophage functions that drive tumor progression. Cancer Res; 77(18); 5129-41. Ā©2017 AACR.
Sigma-Aldrich
Anti-Kindlin-2 Antibody, clone 3A3, clone 3A3, from mouse