Merck
Chemotherapy-associated angiogenesis in neuroblastoma tumors.
The American journal of pathology (2012-01-31)
Martin Michaelis, Nora Hinsch, U Ruth Michaelis, Florian Rothweiler, Thorsten Simon, Hans W ilhelm Doerr, Jaroslav Cinatl, Jindrich Cinatl
ABSTRACT
The influences of cytotoxic drugs on endothelial cells remain incompletely understood. Herein, we examined the effects of chemotherapeutic agents in experimental angiogenesis models and analyzed vessel densities in clinical neuroblastoma tumor samples. Cisplatin (20 to 500 ng/mL), doxorubicin (4 to 100 ng/mL), and vincristine (0.5 to 4 ng/mL), drugs commonly involved in neuroblastoma therapy protocols, induced pro-angiogenic effects in different angiogenesis models. They enhanced endothelial cell tube formation, endothelial cell sprouting from spheroids, formation of tip cells in the sprouting assay, expression of αvβ3 integrin, and vitronectin binding. All three drugs increased global cellular kinase phosphorylation levels, including the angiogenesis-relevant molecules protein kinase Cβ and Akt. Pharmacological inhibition of protein kinase Cβ or Akt upstream of phosphatidylinositol 3-kinase reduced chemotherapy-induced endothelial cell tube formation. Moreover, the investigated chemotherapeutics dose dependently induced vessel formation in the chick chorioallantoic membrane assay. Tumor samples from seven high-risk patients with neuroblastoma were analyzed for vessel density by IHC. Results revealed that neuroblastoma samples taken after chemotherapy consistently showed an enhanced microvessel density compared with the corresponding samples taken before chemotherapy. In conclusion, our data show that chemotherapy can activate endothelial cells by inducing multiple pro-angiogenic signaling pathways and exert pro-angiogenic effects in vitro and in vivo. Moreover, we report a previously unrecognized clinical phenomenon that might, in part, be explained by our experimental observations: chemotherapy-associated enhanced vessel formation in tumors from patients with neuroblastoma.
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, Azide Free, clone LM609, Chemicon®, from mouse