Merck
Impact of dynamin 2 on adenovirus nuclear entry.
Virology (2019-01-21)
Ji Sun Lee, Ashrafali M Ismail, Jeong Yoon Lee, Xiaohong Zhou, Emma C Materne, James Chodosh, Jaya Rajaiya
ABSTRACT
The large GTPase dynamin 2 controls both endosomal fission and microtubule acetylation. Here we report that dynamin 2 alters microtubules and regulates the trafficking of human adenovirus type 37. Dynamin 2 knockdown by siRNA in infected cells resulted in accumulation of acetylated tubulin, repositioning of microtubule organizing centers (MTOCs) closer to cell nuclei, increased virus in the cytosol (with a compensatory decrease in endosomal virus), reduced proinflammatory cytokine induction, and increased binding of virus to the nucleoporin, Nup358. These events led to increased viral DNA nuclear entry and viral replication. Overexpression of dynamin 2 generated opposite effects. Therefore, dynamin 2 inhibits adenovirus replication and promotes innate immune responses by the infected cell. MTOC transposition in dynamin 2 knockdown promotes a closer association with nuclear pore complexes to facilitate viral DNA delivery. Dynamin 2 plays a key role in adenoviral trafficking and influences host responses to infection.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
Bafilomycin A1, Streptomyces griseus, Bafilomycin A1, CAS 88899-55-2, acts as a highly potent and specific inhibitor of vacuolar-type H+-ATPase (Ki = 500 pM). Blocks the fusion of autophagosome with lysosome.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2